Please login to your online account to display your discounted pricing

Promega™ PureYield™ Plasmid Midiprep System


Isolates high-quality plasmid DNA from bacterial cultures

Manufacturer: promega™  A2495

 View more versions of this product

 Disclaimers

U.S. Patent 6,194,562, Australian Patent 740145, Canadian Patent 2,329,067.

Catalog No. PR-A2495

  • / Each
 

Description & Specifications

Specifications

Format Spin or vacuum
Product Type Plasmid Midiprep System
Quantity 100 Preps
Sample Size 50mL
Yield 100 to 200μg
For Use With (Application) Isolates high-quality plasmid DNA from bacterial cultures
Includes 315mL Cell resuspension solution, 315mL Cell lysis solution, 500mL Neutralization solution, 315mL Endotoxin removal wash, 2 × 381mL Column wash, 4 × 25mL Nuclease-free water, 100 PureYield binding columns, 100 PureYield clearing columns
Product Name Plasmid Midiprep System
Storage Requirements Store at 22° to 25°C.
Quantity 100 Preps