Please login to your online account to display your discounted pricing

Promega™ PureYield™ Plasmid Midiprep System


Isolates high-quality plasmid DNA from bacterial cultures

Manufacturer: promega™  A2492

 View more versions of this product

 Disclaimers

U.S. Patent 6,194,562, Australian Patent 740145, Canadian Patent 2,329,067.

Catalog No. PR-A2492

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Isolates high-quality plasmid DNA from bacterial cultures
Format Spin or vacuum
Product Type Plasmid Midiprep System
Sample Size 50mL
Yield 100 to 200μg
Includes 75mL Cell resuspension solution, 75mL Cell lysis solution, 130mL Neutralization solution, 85.3mL Endotoxin removal wash, 210mL Column wash, 25mL Nuclease-free water, 25 PureYield binding columns, 25 PureYield clearing columns
Quantity 25 Preps
Product Name Plasmid Midiprep System
Storage Requirements Store at 22° to 25°C.
Quantity 25 Preps