Please login to your online account to display your discounted pricing
psiCHECK™ Vectors
Provide a quantitative and rapid approach for initial optimization of RNA interference (RNAi)
Manufacturer: promega corportaion  C8011
 View more versions of this product
Catalog No. PR-C8011
  • / Each
Description & Specifications
Specifications
Storage Requirements -20°C
Vector psiCHECK-1 vector
For Use With (Application) Provide a quantitative and rapid approach for initial optimization of RNA interference (RNAi), monitors changes in expression of target gene
Quantity 20μg
Reporter Gene Renilla Luciferase, Firefly Luciferase