Please login to your online account to display your discounted pricing

Promega™ Proteasome-Glo™ Chymotrypsin-Like Cell-Based Assay System


Measures the chymotrypsin-like protease activity of the proteasome complex in cultured cells

Manufacturer: promega™  G8661

 View more versions of this product

Catalog No. PRG8661

  • / Each
 

Description & Specifications

Specifications

Quantity 5 x 10mL
Storage Requirements -20°C
Format Liquid
pH 7.2