Please login to your online account to display your discounted pricing

Promega™ ProFluor™ Tyrosine Phosphatase Assay Systems DFS Item


Measures purified tyrosine phosphatase enzyme activity in a multiwell plate format and involves “add-mix-read” steps only

Manufacturer: promega™  V1280

 View more versions of this product

Catalog No. PR-V1280

  • / Each
 

Description & Specifications

Specifications

Storage Requirements -20°C
Quantity 4 Plates
Product Type Tyrosine Phosphatase assay
Format 96 or 384-well Plate
For Use With (Application) Measures the activity of purified Tyrosine Phosphatases in 96 or 384-well plates, ideal for high throughput applications