Please login to your online account to display your discounted pricing

Promega™ ProFluor™ Src-Family Kinase Assay Systems

Measures the activity of purified Src-family tyrosine kinases (Src, Lck, Lyn, Fyn, and Hck tested) in a multiwell plate format and involves “add-mix-read” steps only

$1,211.24 - $1,211.24

Specifications

Product Type Src Family Protein Kinase assay
Format 96 or 384-well Plate
For Use With (Application) Measures the activity of purified Src-family Tyrosine Kinases in 96 or 384-well plates, ideal for high throughput applications
Storage Requirements -20°C
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

PR-V1270

 
promega™
V1270
4 Plates Each for $1,211.24
Description & Specifications

Specifications

Product Type Src Family Protein Kinase assay
Format 96 or 384-well Plate
For Use With (Application) Measures the activity of purified Src-family Tyrosine Kinases in 96 or 384-well plates, ideal for high throughput applications
Storage Requirements -20°C

  • Ideal for high-throughput applications
  • Assay begins with a standard kinase reaction performed with a provided Src-family kinase bisamide rhodamine 110 peptide substrate fluorescence intensity measured in this assay is inversely correlated with kinase activity
  • A control peptide (AAF-AMC) is included to control for compounds that may inhibit the protease
  • The assay produces excellent Z« values (>0.7) in either 96 or 384-well plate formats and easily distinguishes known Src-family kinase inhibitors from other compounds