missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega ProFluor™ PKA Assay DFS Item

Simple add–mix–read format for higher throughput

$1,211.24 - $1,211.24

Specifications

Product Type PKA assay system
Format 96 or 384-well Plate
For Use With (Application) Measures Protein Kinase A activity in a multiwell plate format using Fluorescence intensity readout
Storage Requirements -20°C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PR-V1240 View Documents Promega™
V1240
4 Plates Each for $1,211.24
Description

Description

•    Measures protein kinase A activity in a multiwell plate format using fluorescence intensity readout
•    Homogeneous assay is performed in a single well
•    Highly predictive Z values are >0.8 in 96– and 384–well plates
•    No radioactive waste or disposal issues

Patent pending.
This product is covered by U.S. Patents 4,557,862 and 4,640,893 and is sold for research use only. All other used, including, but not limited to, use as a clinical diagnostic or therapeutic, require a separate license. Please contact Promega Corporation for details.

Specifications

Specifications

PKA assay system
96 or 384-well Plate
Measures Protein Kinase A activity in a multiwell plate format using Fluorescence intensity readout
-20°C