missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega PKCδ Kinase Enzyme System DFS Item

Recombinant full-length human PKC delta was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Protein kinase C delta is a member of the protein kinase C (PKC) family of serine-threonine kinases.

$615.83 - $615.83

Specifications

Format Fluid
Molecular Weight 104
Storage Requirements <−65°C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRV3401 View Documents Promega™
V3401
10μg Each for $615.83
 
Description

Description

  • Easily Screen and Profile PKCδ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications

Specifications

Fluid
104
<−65°C