Please login to your online account to display your discounted pricing

Promega™ Pgp-Glo™ Assay System DFS Item


Detects effects of compounds on recombinant human P-glycoprotein (Pgp) in a cell membrane fraction

Manufacturer: promega™  V3591

 View more versions of this product

 Disclaimers

1The method of recombinant expression of Coleoptera luciferase is covered by U.S. Pat. Nos. 5,583,024, 5,674,713 and 5,700,673.

Catalog No. PRV3591

  • / Each
 

Description & Specifications

Specifications

Includes 1 x 10mL Pgp Glo Assay Buffer, 1 x 10mL ATP detection Buffer, 1 x 1 vial ATP detection substrate, 1 x 100μL Verapamil, 1 x 1mL MgATP, 1 x 250μL Sodium Vanadate
pH 6
Quantity 10mL
Storage Requirements -70°C
Format Fluid