Please login to your online account to display your discounted pricing

Promega™ pGL4 Luciferase Reporter Vectors


Available in numerous configurations for enhanced usability and convenience

Manufacturer: promega™  E6651

 View more versions of this product

Catalog No. PRE6651

  • / Each
 

Description & Specifications

Specifications

Storage Requirements -20°C
Quantity 20μg
Vector pGL4.10[luc2] vector
For Use With (Application) Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis
Primer RV primer 3 and RV primer 4
Concentration 1μg/μL
Storage Buffer 10mM Tris HCl (pH 7.4), 1mM EDTA
Reporter Gene luc2A
Size 20μg