Please login to your online account to display your discounted pricing

Promega™ pGL4 Luciferase Reporter Vectors


Available in numerous configurations for enhanced usability and convenience

Manufacturer: promega™  E6921

 View more versions of this product

Catalog No. PRE6921

  • / Each
 

Description & Specifications

Specifications

Promoter HSV-TK
Quantity 20μg
For Use With (Application) Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis
Primer Primer 3 and primer 4
Concentration 1μg/μL
Reporter Gene hRlucB
Vector pGL4.74[hRluc/TK] vector
Storage Requirements -20°C
Storage Buffer 10mM Tris HCl (pH 7.4), 1mM EDTA
Size 20μg