Please login to your online account to display your discounted pricing

Promega™ pGL4 Luciferase Reporter Vectors DFS Item


Available in numerous configurations for enhanced usability and convenience

Manufacturer: promega™  E6911

 View more versions of this product

Catalog No. PRE6911

  • / Each
 

Description & Specifications

Specifications

Promoter SV40
Storage Requirements -20°C
For Use With (Application) Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis
Vector pGL4.73[hRluc/SV40] vector
Primer Primer 3 and primer 4
Reporter Gene hRlucB
Storage Buffer 10mM Tris HCl (pH 7.4), 1mM EDTA
Concentration 1μg/μL
Quantity 20μg