Please login to your online account to display your discounted pricing

Promega™ pGL4 Luciferase Reporter Vectors


Available in numerous configurations for enhanced usability and convenience

Manufacturer: promega™  E8411

 View more versions of this product

Catalog No. PRE8411

  • / Each
 

Description & Specifications

Specifications

Concentration 1μg/μL
Quantity 20μg
Storage Buffer 10mM Tris HCl (pH 7.4), 1mM EDTA
Primer RV primer 3 and RV primer 4
Storage Requirements -20°C
Vector pGL4.23[luc2/minP] vector
For Use With (Application) Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis
Promoter minP
Reporter Gene luc2A