Please login to your online account to display your discounted pricing

Promega™ pGL4 Luciferase Reporter Vectors DFS Item


Available in numerous configurations for enhanced usability and convenience

Manufacturer: promega™  E1370

 View more versions of this product

Catalog No. PRE1370

  • / Each
 

Description & Specifications

Specifications

Concentration 1μg/μL
Reporter Gene luc2A
Quantity 20μg
Storage Buffer 10mM Tris HCl, 1mM EDTA (pH 7.4 at 25°C)
For Use With (Application) Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis
Marker Gene Hygro
Vector pGL4.35[luc2P/9XGAL4UAS/Hygro] vector
Storage Requirements -20°C