Please login to your online account to display your discounted pricing

Promega™ pGL4 Luciferase Reporter Vectors


Available in numerous configurations for enhanced usability and convenience

Manufacturer: promega™  E8491

 View more versions of this product

Catalog No. PR-E8491

  • / Each
 

Description & Specifications

Specifications

Vector pGL4.32[luc2P/NF-κB-RE/Hygro] vector
Marker Gene Hygro
Storage Requirements -20°C
Reporter Gene luc2A
For Use With (Application) Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis
Protein Degradation Sequence hPEST
Primer RV primer 3 and RV primer 4
Quantity 20μg
Concentration 1μg/μL
Promoter NF-κB-RE
Storage Buffer 10mM Tris HCl (pH 7.4), 1mM EDTA