Please login to your online account to display your discounted pricing

Promega™ pGEM™ Vectors DFS Item


Enable blue/white screening and contain both the SP6 and T7 RNA polymerase promoters

Manufacturer: promega™  P2411

 View more versions of this product

Catalog No. PR-P2411

  • / Each
 

Description & Specifications

Specifications

Includes 500μL Glycerol stock of bacterial strain JM109
Restriction Site SfiI, SacI, EcoRI, SalI, XhoI, BamHI, ApaI, XbaI, NotI, SphI, NsiI and HindIII
Vector pGEM-11Zf(+) vector
Promoter Sp6 and T7
Quantity 20μg
Storage Requirements Vector at -20°C and bacterial strain at -70°C
For Use With (Application) Synthesis of RNA in-vitro and allows selection of recombinants by blue/white screening