Please login to your online account to display your discounted pricing

Promega™ pGEM™-T and pGEM™-T Easy Vector Systems


Convenient systems for the routine subcloning of PCR products

Manufacturer: promega™  A1380

 View more versions of this product

Catalog No. PR-A1380

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) For the routine subcloning of PCR products
Storage Requirements Store competent cells at -70°C, store all other components at -20°C
Vector pGEM-T easy vector II
Promoter Sp6 and T7
Includes 1.2μg pGEM-T easy vector (50ng/μL), 12μL control insert DNA (4ng/μL), 200μL rapid ligation Buffer (2X), 100U T4 DNA Ligase, JM109 1.2mL competent cells high efficiency (6 x 200μL)
Restriction Site BstZI, NotI and EcoRI
Quantity 20 Reactions