Please login to your online account to display your discounted pricing

Promega™ pGEM™-T and pGEM™-T Easy Vector Systems


Convenient systems for the routine subcloning of PCR products

Manufacturer: promega™  A1360

 View more versions of this product

Catalog No. PR-A1360

  • / Each
 

Description & Specifications

Specifications

Quantity 20 Reactions
Promoter Sp6 and T7
For Use With (Application) For the routine subcloning of PCR products
Storage Requirements -20°C
Includes 1.2μg pGEM-T easy vector (50ng/μL), 12μL control insert DNA (4ng/μL), 200μL rapid ligation Buffer (2X), 100U T4 DNA Ligase
Restriction Site BstZI, NotI and EcoRI
Vector pGEM-T easy vector I