Please login to your online account to display your discounted pricing

Promega™ PepTag™ Nonradioactive Protein Kinase Assays


Rapid, sensitive screening of column fractions or for kinase activators and inhibitors

Manufacturer: promega™  V5340

 View more versions of this product

Catalog No. PR-V5340

  • / Each
 

Description & Specifications

Specifications

Quantity 120 Reactions
For Use With (Application) Uses brightly colored Fluorescent Peptide Substrates to detect less than 2ng of purified Kinase in under two Hours
Storage Requirements -70°C
Product Type Non radioactive cAMP-dependent assay
Includes Purified Kinase as a positive control, 650μL PepTag™ PKA Reaction 5X Buffer, 650μL PKA Activato r 5X Solution, 25μL cAMP-dependent Protein Kinase, Catalytic Su