Promega™ PepTag™ Nonradioactive Protein Kinase Assays DFS Item
The PepTag Non-Radioactive Protein Kinase Assay Systems provide a rapid, sensitive, non-radioactive method to detect either Protein Kinase C (PKC) or Protein Kinase A (PKA) activity
Manufacturer:  Promega™ V5330
 View more versions of this product
Catalog No. PR-V5330
Includes: Purified kinase as a positive control
Description
Description
  • Use brightly colored fluorescent peptide substrates that are highly specific for PKC and PKA
  • Detect less than 2ng of purified kinase in under two hours
  • Can detect kinase activity in partially purified samples as well as in purified preparations of enzymes
  • Low background
  • Quantitation of phosphorylated peptide can be accomplished using a densitometer, spectrophotometer, 96-well plate reader or fluorometer
U.S. Patents 5,580,747 and other patents pending.
Specifications
Specifications
Non radioactive PKC assay
120 Reactions
Uses brightly colored Fluorescent Peptide Substrates to detect less than 2ng of purified Kinase in under two Hours
<−65°C