Please login to your online account to display your discounted pricing

Promega™ PepTag™ Nonradioactive Protein Kinase Assays

Rapid, sensitive screening of column fractions or for kinase activators and inhibitors

$513.60 - $513.60

Specifications

Quantity 120 Reactions
For Use With (Application) Uses brightly colored Fluorescent Peptide Substrates to detect less than 2ng of purified Kinase in under two Hours
Storage Requirements -70°C
View More Specs
Includes:  Purified kinase as a positive control
Products
Catalog Number Mfr. No. Product Type Price Quantity    

PR-V5330

 
promega™
V5330
Non radioactive PKC assay Each for $513.60

PR-V5340

 
promega™
V5340
Non radioactive cAMP-dependent assay Each for $513.60
Description & Specifications

Specifications

Quantity 120 Reactions
For Use With (Application) Uses brightly colored Fluorescent Peptide Substrates to detect less than 2ng of purified Kinase in under two Hours
Storage Requirements -70°C

  • Use brightly colored fluorescent peptide substrates that are highly specific for PKC and PKA
  • Detect less than 2ng of purified kinase in under two hours
  • Can detect kinase activity in partially purified samples as well as in purified preparations of enzymes
  • Low background
  • Quantitation of phosphorylated peptide can be accomplished using a densitometer, spectrophotometer, 96-well plate reader or fluorometer

U.S. Patents 5,580,747 and other patents pending.