Promega™ PDE-Glo™ Phosphodiesterase Assay DFS Item
Luminescent, high-throughput screening method for measuring cyclic nucleotide phosphodiesterase (PDE) activity from purified sources
$884.88 - $884.88
Specifications
Storage Requirements -20°C
View More Specs
Disclaimers:
Patent pending.
The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Quantity Price Quantity  
Catalog Number Mfr. No. Includes Quantity Price Quantity  
PRV1361 Promega™
V1361
1 x 1.2mL 5X PDE-Glo™ Reaction Buffer, 1 x 600μL 5X PDE-Glo™ detection Buffer, 1 x 20μL Protein Kinase A, 1 x 600μL 5X PDEGlo Termination Buffer, 1 x 500μL 1000 Assays Each for $884.88
Specifications
Description
Cyclic nucleotide PDEs are involved in countless cellular processes due to their ability to hydrolyze, and therefore control, the levels of the second-messenger signaling molecules cAMP and cGMP. The availability of selective inhibitors for PDEs has facilitated their use as tools to study cyclic nucleotide signaling and paved the way to investigate the role of PDEs in cellular and tissue pathologies.
  • Allows lead candidates to be identified from compound libraries
  • Optimized to work with both cAMP- and cGMP-dependent PDEs
  • Requires less than one hour from start to finish once the PDE reaction is complete
  • Designed for 384-well plates, but can easily be scaled for 96- or 1536-well plates
Description & Specifications
Specifications
-20°C