Promega™ PCR Master Mix DFS Item
Premixed 2X solution of Taq DNA Polymerase, dNTPs and Reaction Buffer.
Manufacturer:  Promega™ M7501
 View more versions of this product
Catalog No. PR-M7501
Description
Description
  • Contains Taq DNA Polymerase, dNTPs, MgCl2, and reaction buffers at optimal concentrations
  • Optimized for use in routine PCR with templates in the range of 0.2 to 2kb
  • Amplifies as little as two copies of target template
  • Scalable for 10, 25, or 50μL reactions
Specifications
Specifications
PCR, RT-PCR
10.0
−30 to −10°C
2X
1 x 250μL PCR master mix, 2X, 1 x 1250μL Nuclease free Water, Taq DNA Polymerase, dNTPs, MgCl2 and reaction Buffers
10 Reactions
50μL