Learn More
Promega™ P450-Glo™ CYP450 Assay Systems
Provide a complete set of reagents for performing luminescent cytochrome P450 assays
Manufacturer: Promega™ V8812
Includes: Luciferin Detection Reagent; P450-Glo Buffer; 5mM Luciferin-CEE, -ME, -H, -BE, -PFBE, -PPXE, or IPA
Description
The assays are designed to measure the activities of P450s from recombinant and native sources and for testing the effects of analytes such as drugs and new chemical entities on P450 activities. These luminescent assays exhibit high sensitivity, low background signals and broad dynamic range.
The assays employ luminogenic P450 substrates that are derivatives of beetle luciferin, a substrate for luciferase enzymes. The derivatives are not substrates for luciferase but are converted by P450s to luciferin, which in turn reacts with luciferase to produce light that is directly proportional to the activity of the P450.
- Measure the activity of P450s from recombinant or native fractions
- Test the effects of analytes including drugs and new chemical entities on P450 activities
- Simple “add-and-read” procedures feature high sensitivity, low background signals, and broad dynamic range
- Scalable to 384-well format
- Eliminate interference from fluorescent test compounds
- Exhibit “glow-type” signal with a half-life of over two hours, eliminating the need for luminometers with injectors and allowing for batch plate processing
- Formulation reduces the incidence of false positives
- CYP3A4 Assay with Luciferin-IPA contains a substrate for cytochrome 3A4 that is very well suited for all applications involving human CYP3A4 and is well suited for cell-based applications
Screen drugs and new chemical entities for their capacity to modulate CYP450 activities in native or recombinant fractions; Measure recombinant CYP450 activities in membrane fractions from heterologous expression systems, such as Sf9 cells and E. coli; and in microsomal fractions from tissues
Specifications
50mL | |
Homogeneous, Luminescent | |
For measuring cytochrome P450 activity |
−30 to −10°C | |
1 x 1 Each Luciferin detection reagent, 1 x 50mL Reconstitution Buffer, 1 x 750μL Luciferin-BE | |
CYP3A7 Assay |
Promotions
-
Spend More, Get More on Promega Products
Purchase up to $1,000 in Promega reagents and get a matching amount of products at no additional cost.: 06/30/21
U.S. Patent 6,602,677; Australian Pat. No. 754312 and other patents pending.
The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713 and 5,700,673.