Please login to your online account to display your discounted pricing

Promega™ P450-Glo™ CYP450 Assay Systems


Provide homogeneous, luminescent methods for measuring cytochrome P450 activity

Manufacturer: promega™  V8801

 View more versions of this product

 Disclaimers

U.S. Patent 6,602,677; Australian Pat. No. 754312 and other patents pending.
The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713 and 5,700,673.

Catalog No. PR-V8801

  • / Each
 

Description & Specifications

Specifications

Quantity 10mL
For Use With (Application) For measuring cytochrome P450 activity
Product Type P450-Glo CYP3A4 Assay
Storage Requirements -20° to -70°C
Detection Method Homogeneous, Luminescent
Kit Contents 1 x 1 Each Luciferin detection reagent, 1 x 10mL Reconstitution Buffer, 1 x 100μL Luciferin-BE