Please login to your online account to display your discounted pricing

Promega™ ONE-Glo™ Luciferase Assay System DFS Item


Provides a highly sensitive, robust, homogeneous assay for detection of firefly luciferase reporter gene expression in mammalian cells

Manufacturer: promega™  E6110

 View more versions of this product

 Disclaimers

Patent pending.
The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
Certain applications of this product may require licenses from others.

Catalog No. PRE6110

  • / Each
 

Description & Specifications

Specifications

Storage Requirements -20°C
pH 6.7
For Use With (Application) Detection of firefly luciferase reporter gene expression in mammalian cells
Quantity 10mL
Detection Method Luminescence
For Use With (Equipment) 384- and 1536-well plates
Includes 1 x 10mL ONE-Glo™ Luciferase Assay Buffer, 1 x 1 vial ONE-Glo™ Luciferase assay substrate