Please login to your online account to display your discounted pricing

Promega™ ONE-Glo™ Assay


A reagent that is more robust and stable when reconstituted, and less aromatic than
standard reagents created with luciferin

Manufacturer: promega™  E7120

 View more versions of this product

Catalog No. PRE7120

  • / Each
 

Description & Specifications

Specifications

pH 7.5
Quantity 10 Plates
Format Fluid
Storage Requirements -20°C
Kit Contents 1 x 100mL ONE-Glo™ Luciferase Assay Buffer, 1 x 1 Vial ONE-Glo™ Luciferase assay substrate, 2 x 50μL GF-AFC substrate, 1 x 50mL Assay Buffer