Please login to your online account to display your discounted pricing

Promega™ Nonradioactive Phosphatase Assay Systems


Fast, flexible alternative for measuring protein phosphatase activity

Manufacturer: promega™  V2471

 View more versions of this product

Catalog No. PR-V2471

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 4°C
Product Type Tyrosine system
Quantity 96 Reactions
For Use With (Application) Can be used with partially fractionated and purified samples as well as in crude cell or tissue extracts