Promega NanoLuc™ pNL2 Vectors
Configured with NanoLuc Luciferase for use in reporter gene assays of transcriptional control—ideal for cloning of known or putative promoter region and establishment of stable cell line through Hygromycin selection
Supplier: Promega N1061
Description
- Clone a Putative Promoter for Expression in Mammalian Cells and Select Stable Transfectants
- Offers two engineered genes for your reporter assay: Nluc and NlucP
- NanoLuc(R) enzyme and its substrate are about 100-fold brighter than other luciferases, producing a high intensity, glow-type luminescence
- Use for transient transfection or select for stable expression with hygromycin
Specifications
pNL2.1(Nluc/Hygro) | |
-30°C to -10°C | |
Liquid |
8 | |
20 μg |