Please login to your online account to display your discounted pricing

Promega™ MgCl2 Solution, 25mM DFS Item

For PCR applications.

$46.01 - $65.27

Specifications

Concentration 25mM
For Use With (Application) Efficiently reverse transcribes RNA into cDNA
Storage Requirements -20°C
View More Specs
Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

PR-A3511

DFS Item
 
promega™
A3511
2 x 750μL MgCl2 1.5mL Each for $46.01

PR-A3513

DFS Item
 
promega™
A3513
25mL of MgCl2 25mL Each for $65.27
Description & Specifications

Specifications

Concentration 25mM
For Use With (Application) Efficiently reverse transcribes RNA into cDNA
Storage Requirements -20°C

  • AMV Reverse Transcriptase synthesizes single-stranded cDNA from total or poly(A)+ RNA Both Oligo(dT)15 and Random Primers are included, allowing cDNA synthesis from virtually any RNA source
  • A polyadenylated 1.2kb RNA transcript is provided as a control template