Promega™ Maxwell™ 16 RNA Purification Kits DFS Item
Use with the Maxwell 16 instrument to provide high yields of DNA-free RNA
Manufacturer:  Promega™ AS1220
 View more versions of this product
Catalog No. PRAS1220
Disclaimers:
For Laboratory Use.
Specifications Description & Specifications
Specifications
Automated total RNA purification, qRT-PCR, RT-PCR and cDNA synthesis
Maxwell™ 16 LEV total RNA cartridges 48, RNA Lysis Buffer (RLA) 50mL, RNA dilution Buffer (RDB) 44mL, 97.4% beta-mercaptoethanol (BME) 0.9mL, clearing agent (CAA) 6.5mL, Nuclease-free Water 25mL, clearing columns 50, collection tubes 50,
48 Preps
Promotions
Promotions
  • Purchase up to $1,000 in any combination of Promega™ reagents and receive an equivalent amount of complimentary Promega products of your choice. This is an NLSU exclusive offer. You must apply and qualify for the program to be eligible.
    06/30/18