Promega Maxwell™ 16 RNA Purification Kits
Use with the Maxwell 16 instrument to provide high yields of DNA-free RNA
$839.69 - $839.69
Specifications
For Use With (Application) | Automated total RNA purification, qRT-PCR, RT-PCR and cDNA synthesis |
---|---|
Quantity | 48 Preps |
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
PRAS1220
|
Promega
AS1220 |
Maxwell™ 16 LEV total RNA cartridges 48, RNA Lysis Buffer (RLA) 50mL, RNA dilution Buffer (RDB) 44mL, 97.4% beta-mercaptoethanol (BME) 0.9mL, clearing agent (CAA) 6.5mL, Nuclease-free Water 25mL, clearing columns 50, collection tubes 50, |
Each for $839.69
|
|
|||||
Description
- No detectable contaminating genomic DNA means fewer repeated experiments or unexplained variable results
- Efficient processing, superior sample capacity
- For most sample types, eluted RNA concentration is directly ready for QRT-PCR; no post-purification concentration required
Maxwell 16 Total RNA Purification Kit
- Extracts total RNA from white blood cell fraction of whole blood, tissue culture cell lines
- PAXgene™-stabilized whole blood and plant leaf tissue can be used in RNA cleanup applications from TRIZOL™ extractions or in vitro transcription reactions
- β-Mercaptoethanol is included for use with certain sample types
- Provide high-concentration (100ng/μL), essentially DNA-free total RNA from cultured cells, mammalian tissue and PAXgene-stabilized white blood cells
- It's personal automation working for you, so you can get more consistent gene expression analysis results with less hands-on labor
Specifications
Automated total RNA purification, qRT-PCR, RT-PCR and cDNA synthesis | |
48 Preps |
For Research Use Only (RUO)