Please login to your online account to display your discounted pricing

Promega™ Maxwell™ 16 Purification Kits


Used with Maxwell 16 Instruments to provide an easy method for efficient, automated purification of genomic DNA

Manufacturer: promega™  AS1140

 View more versions of this product

Maxwell 16 DNA Purification Instrument (Mfr. No. AS1120 not for use with Maxwell 16 MDx); LEV Kits also require the LEV Hardware Kit (Mfr. No. AS1250)

Catalog No. PR-AS1140

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Automated extraction of genomic DNA from mammalian cells
Quantity 48 Preps
Storage Requirements Store at room temperature.
Product Type Cell LEV DNA Kit
Includes 48 Maxwell™ 16 LEV cartridges(MCB), 50 LEV purification plungers, 50 Elution tubes, 20mL Elution Buffer
For Use With (Equipment) Maxwell™ 16 and 16 MDx instruments