Please login to your online account to display your discounted pricing

Promega™ Maxwell™ 16 Purification Kits


Used with Maxwell 16 Instruments to provide an easy method for efficient, automated purification of genomic DNA

Manufacturer: promega™  AS1290

 View more versions of this product

Maxwell 16 DNA Purification Instrument (Mfr. No. AS1120 not for use with Maxwell 16 MDx); LEV Kits also require the LEV Hardware Kit (Mfr. No. AS1250)

Catalog No. PRAS1290

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Automated genomic DNA extraction from whole blood samples collected in EDTA, ACD or heparin tubes
Size 300μL
For Use With (Equipment) Maxwell™ 16 and 16 MDx instruments
Includes 48 Maxwell™ 16 MCD LEV cartridges, 50 LEV purification plungers, 50 Elution tubes, 20mL Lysis Buffer, two 1mL Proteinase K solution, 20mL Elution Buffer
No. of Reactions 48 Preps
Product Type LEV Blood DNA Kit
Quantity 300μL
Product Name LEV Blood DNA Kit
Storage Requirements Store at room temperature.
Quantity 48 Preps