Please login to your online account to display your discounted pricing

Promega™ Maxwell™ 16 Purification Kits


Used with Maxwell 16 Instruments to provide an easy method for efficient, automated purification of genomic DNA

Manufacturer: promega™  AS1150

 View more versions of this product

Maxwell 16 DNA Purification Instrument (Mfr. No. AS1120 not for use with Maxwell 16 MDx); LEV Kits also require the LEV Hardware Kit (Mfr. No. AS1250)

Catalog No. PRAS1150

  • / Each
 

Description & Specifications

Specifications

Product Type Viral Total Nucleic Acid Kit
Includes 48 Maxwell™ 16 MCC LEV cartridges, 50 LEV purification plungers, 50 Elution tubes, 20mL Lysis Buffer, two 1mL Proteinase K solution, 20mL Nuclease-free Water
No. of Reactions 48 Preps
Storage Requirements Store at room temperature.
Quantity 100, 200, or 300μL
For Use With (Equipment) Maxwell™ 16 and 16 MDx instruments
For Use With (Application) Automated extraction of viral total nucleic acid from serum, plasma and other sample types