Please login to your online account to display your discounted pricing

Promega™ Maxwell™ 16 Purification Kits DFS Item


Used with Maxwell 16 Instruments to provide an easy method for efficient, automated purification of genomic DNA

Manufacturer: promega™  AS1030

 View more versions of this product

Maxwell 16 DNA Purification Instrument (Mfr. No. AS1120 not for use with Maxwell 16 MDx); LEV Kits also require the LEV Hardware Kit (Mfr. No. AS1250)

Catalog No. PR-AS1030

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) Maxwell™ 16 and 16 MDx instruments
Product Type Tissue DNA Kit
Quantity 50mg
Storage Requirements Store at room temperature.
Includes 48 Maxwell™ 16 tissue DNA cartridges, 50 purification plungers, 50 Elution tubes, 20mL Elution Buffer
For Use With (Application) Automated DNA purification from tissue samples
No. of Reactions 48 Preps