Learn More
Promega Maxwell™ 16 Purification Kits
Used with Maxwell 16 Instruments to provide an easy method for efficient, automated purification of genomic DNA
Supplier: Promega AS1020
Includes:
All kits include 48 Maxwell kit-specific cartridges, 50 purification plungers, 50 elution tubes, and 20mL elution buffer. FFPE Tissue LEV DNA Purification Kit also includes 32mL lysis buffer, 2 x 10mg proteinase K, and 15mL incubation buffer. Viral Total Nucleic Acid Kit also includes 20mL lysis buffer, 2 x 1mL proteinase K solution, and 20mL nuclease-free water. LEV Blood DNA Kit also includes 20mL lysis buffer and 2 x 1mL proteinase K solution.
Requires : Maxwell 16 DNA Purification Instrument (Mfr. No. AS1120 not for use with Maxwell 16 MDx); LEV Kits also require the LEV Hardware Kit (Mfr. No. AS1250)
Description
- Automated DNA Extraction from Cells
- Compatible with cells grown in suspension or attached to plates
- Processes sample volumes up to 400μL
- For use with the Maxwell(R) 16 Instrument
U.S. Patents 6,027,945 and 6,368,800; Australian Pat. No. 732756.
Mfr. Nos. AS1010, AS1020, AS1030, and AS1140: This product is licensed for use under U.S. Patent 6,174,704.
Mfr. Nos. AS1130, AS1140: U.S. Patent 6,673,631.
Mfr. Nos. AS1130, AS1140: Patent pending.
Specifications
15°C to 30°C | |
Automated genomic DNA purification from cell and tissue culture, eukaryotic and prokaryotic cell samples | |
48 Preps |
Maxwell™ 16 and 16 MDx instruments | |
48 Maxwell™ 16 cell DNA cartridges, 50 purification plungers, 50 Elution tubes, 20mL Elution Buffer | |
Cell DNA Kit |