Please login to your online account to display your discounted pricing

Promega™ Maxwell™ 16 Purification Kits


Used with Maxwell 16 Instruments to provide an easy method for efficient, automated purification of genomic DNA

Manufacturer: promega™  AS1010

 View more versions of this product

Maxwell 16 DNA Purification Instrument (Mfr. No. AS1120 not for use with Maxwell 16 MDx); LEV Kits also require the LEV Hardware Kit (Mfr. No. AS1250)

Catalog No. PR-AS1010

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at room temperature.
Product Type Blood DNA Kit
Includes 48 Maxwell™ 16 Blood DNA cartridges, 50 purification plungers, 50 Elution tubes, 20mL Elution Buffer
For Use With (Equipment) Maxwell™ 16 and 16 MDx instruments
No. of Reactions 48 Preps
For Use With (Application) Automated genomic DNA purification from whole blood and buffy coat samples
Quantity 400μL