missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega MAO-Glo™ Monoamine Oxidase Assay System
The MAO-Glo™ Assay provides a homogeneous luminescent method for measuring monoamine oxidase activity from recombinant and native sources and for testing the effects of test compounds on MAO activity.
$1076.60 - $3128.45
Specifications
For Use With (Application) | Provides a homogeneous luminescent method for measuring MAO activity from recombinant and native sources |
---|---|
Content And Storage | −30 to −10°C |
Catalog Number | Mfr. No. | Product Type | Quantity | Content And Storage | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Quantity | Content And Storage | Price | Quantity | |||
PR-V1401
|
Promega
V1401 |
Monoamine Oxidase assay system | 200 Assays | −30 to −10°C |
Each for $1,076.60
|
|
|||
PR-V1402
|
Promega
V1402 |
Assay System | 1000 Assays | −30 to −10°C |
Each for $3,128.45
|
|
|||
Description
- Sensitive Detection of Monoamine Oxidase (MAO) Activity
- Luminescence-based method eliminates need for time-consuming analyses such as HPLC
- Simple add and read protocol ideal for high-throughput screening
- Less MAO enzyme required than typical HPLC or fluorometric methods
Specifications
Provides a homogeneous luminescent method for measuring MAO activity from recombinant and native sources | |
−30 to −10°C |