Promega MagneGST™ Protein Purification System
MagneGST Protein Purification System provides a simple, rapid and reliable method for the purification of glutathione-S-transferase (GST) fusion proteins.
Supplier: Promega V8600
Description
- Simple Method for Glutathione-S-Transferase Fusion Protein Purification
- One-step purification of multiple samples; no need for high-speed centrifugation after cell lysis
- Use with 1-50mL of cell culture
- Achieve high yields with little to no nonspecific background
Specifications
4mL MagneGST™ Glutathione particles, 2mL Glutathione, 80mL MagneGST™ binding/Wash Buffer, 12mL MagneGST™ cell lysis reagent, 1000μL RQ1 RNase-free DNase, 2mL Tris Buffer, 0.5M (pH 8.1) | |
40 Reactions | |
Magnetic Bead Protein Purification System |
COMPONENTS AT DIFFERENT TEMPS | |
Protein purification of Glutathione-S-transferase fusion Proteins from cleared or crude E. coli cell lysate, high-throughput purification of GST-tagged Proteins using auto mated platforms |