Learn More
Promega MagneGST™ Protein Purification System
MagneGST Protein Purification System provides a simple, rapid and reliable method for the purification of glutathione-S-transferase (GST) fusion proteins.
$376.41 - $376.41
Specifications
Content And Storage | COMPONENTS AT DIFFERENT TEMPS |
---|---|
For Use With (Application) | Protein purification of Glutathione-S-transferase fusion Proteins from cleared or crude E. coli cell lysate, high-throughput purification of GST-tagged Proteins using auto mated platforms |
Type | Protein Purification System |
Includes: MagneGST Glutathione Particles (also available separately), (4mL); Glutathione, (2mL); MagneGST Binding/Wash Buffer, (80mL); MagneGST Cell Lysis Reagent, (12mL); RQ1 RNase-free DNase, (1000μL); Tris Buffer, 0.5M (pH 8.1), (2mL)
Catalog Number | Mfr. No. | Type | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Quantity | Price | Quantity | ||||
PR-V8600
![]() |
Promega
V8600 |
Protein Purification System | 40 Reactions |
Each for $376.41
|
|
Description
- Simple Method for Glutathione-S-Transferase Fusion Protein Purification
- One-step purification of multiple samples; no need for high-speed centrifugation after cell lysis
- Use with 1-50mL of cell culture
- Achieve high yields with little to no nonspecific background
This product or portions thereof is manufactured under license from Amrad Corporation Limited. For non-commercial research use only. All other uses require a license from Amrad Corporation Limited, Unit 6, 663 Victoria Street, Abbotsford, Victoria 3067, Australia, under U.S. Patent 5,654,176, Australian Patent 607511, Canadian Patent 1338903 and other issued patents.
Patent pending.
This product is licensed for use under U.S. Patent 6,174,704.
Certain applications of this product are covered by patents issued and applicable in certain countries. Because purchase of this product does not include a license to perform any patented application, users of this product may be required to obtain a patent license depending upon the particular application and country in which the product is used.
Specifications
COMPONENTS AT DIFFERENT TEMPS | |
Protein purification of Glutathione-S-transferase fusion Proteins from cleared or crude E. coli cell lysate, high-throughput purification of GST-tagged Proteins using auto mated platforms | |
Protein Purification System |