Please login to your online account to display your discounted pricing
Promega™ Gutathione Particles for MagneGST™ Protein Purification System DFS Item
Manufacturer: promega™  V8611
 View more versions of this product
Catalog No. PR-V8611
  • / Each
Description & Specifications
Specifications
Quantity 4mL
Storage Requirements 4° or -70°C
For Use With (Application) Protein purification of Glutathione-S-transferase fusion Proteins from cleared or crude E. coli cell lysate, high-throughput purification of GST-tagged Proteins using auto mated platforms