Please login to your online account to display your discounted pricing

Promega™ Magne™ Protein G Magnetic Affinity Beads


Allow rapid and efficient capture of antibodies either with manually processed samples or in high-throughput manner

Manufacturer: promega™  G7472

 View more versions of this product

Catalog No. PRG7472

  • / Each
 

Description & Specifications

Specifications

Quantity 5mL
Storage Requirements 2° to 10°C
Particle Size 30 to 80μm
Product Type Magnetic Affinity Bead
Binding Property ≤25mg/mL
Formulation 20% Slurry in 20% Ethanol
For Use With (Application) Antibody purification from Serum, ascites and cell-media samples, auto mated, high-throughput Antibody purification, Antibody purification and concentration from small-volume or dilute source material