missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega Magne™ HaloTag™ Magnetic Affinity Beads DFS Item

The Magne HaloTag™ Beads provide a convenient method to covalently capture HaloTag™ fusion proteins with magnetic particles for protein pull-downs and purification. The beads offer high binding capacity with low nonspecific protein binding.

$165.59 - $165.59

Specifications

Product Type Beads
Formulation 20% Slurry in 10 to 15% Ethanol
For Use With (Application) Auto mated and manual purification of HaloTag™ fusion Proteins
Particle Size 50 to 80μm
Storage Requirements 2 to 10°C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRG7281 View Documents Promega™
G7281
1mL Each for $165.59
 
Description

Description

  • Magnetic Capture of HaloTag(R) Fusion Proteins for Pull-Downs and Purification
  • Bind ≥20mg of purified HaloTag(R) fusion protein per mL of particles
  • Manual or high-throughput applications
Specifications

Specifications

Beads
Auto mated and manual purification of HaloTag™ fusion Proteins
2 to 10°C
20% Slurry in 10 to 15% Ethanol
50 to 80μm