Please login to your online account to display your discounted pricing

Promega™ M-MLV Reverse Transcriptase


RNA-dependent DNA polymerase can be used in primer extension or cDNA synthesis with >5kb mRNA templates

Manufacturer: promega™  M5313

 View more versions of this product

Catalog No. PR-M5313

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) First-strand cDNA synthesis, primer extension
Includes 2 x 1mL Buffer pack
Concentration 5X
Format Liquid
Source Recombinant E. coli Strain
Storage Buffer Tris-HCl (pH 7.5 at 25°C) 20mM, NaCl 0.2M, EDTA 0.1mM, DTT 1mM, 0.01% Nonidet™ P40™ and Glycerol 50%
Storage Requirements -20°C
pH 8.3
Product Name M-MLV Reverse Transcriptase Buffer Pack
Quantity 2 x 1mL