Please login to your online account to display your discounted pricing
Promega™ Luminogenic Enzyme Substrates DFS Item
The pro-luciferin substrates can be used to monitor the activity of specific isoforms of cytochrome P450 or NAT2 as indicated in the name of the substrate
Manufacturer: promega™  P1731
 View more versions of this product
Catalog No. PRP1731
  • / Each
Description & Specifications
Specifications
Substrate Quantity 3mg
Substrate Type Luciferin MultiCYP
For Use With (Application) Monitor the activity of specific isoforms of cytochrome P450 or NAT2