Please login to your online account to display your discounted pricing

Promega™ Luciferase Assay Systems DFS Item


Extremely sensitive and rapid systems for the quantification of firefly luciferase

Manufacturer: promega™  E1500

 View more versions of this product

Catalog No. PR-E1500

  • / Each
 

Description & Specifications

Specifications

Quantity 100 Assays
Format Liquid
Product Type Luciferase Assay System
Includes 1 vial Luciferase Assay Substrate, 10mL Luciferase Assay Buffer, 30mL Cell Culture Lysis 5X Reagent
Storage Requirements -20°C
pH 4.6