Please login to your online account to display your discounted pricing

Promega™ Luciferase Assay Systems DFS Item


Extremely sensitive and rapid systems for the quantification of firefly luciferase

Manufacturer: promega™  E1501

 View more versions of this product

Catalog No. PR-E1501

  • / Pack of 10 Each
 

Description & Specifications

Specifications

pH 4.6
Product Type Luciferase Assay System, 10-pack
Includes 10 × 1 vial Luciferase Assay Substrate, 10 × 10mL Luciferase Assay Buffer
Format Liquid
Quantity 1000 Assays
Storage Requirements -20°C