Please login to your online account to display your discounted pricing

Promega™ Luciferase Assay Systems DFS Item


Extremely sensitive and rapid systems for the quantification of firefly luciferase

Manufacturer: promega™  E4550

 View more versions of this product

Catalog No. PR-E4550

  • / Each
 

Description & Specifications

Specifications

Format Liquid
pH 4.6
Storage Requirements -20°C
Product Type Luciferase Assay System
Includes 1 vial Luciferase Assay Substrate, 105mL Luciferase Assay Buffer
Quantity 1000 Assays