Please login to your online account to display your discounted pricing

Promega™ Luciferase Assay Systems


Extremely sensitive and rapid systems for the quantification of firefly luciferase

Manufacturer: promega™  E4030

 View more versions of this product

Catalog No. PR-E4030

  • / Each
 

Description & Specifications

Specifications

Quantity 100 Assays
Format Liquid
pH 4.6
Includes 1 vial Luciferase Assay Substrate, 10mL Luciferase Assay Buffer, 30mL Reporter Lysis 5X Buffer
Product Type Luciferase Assay System w/Reporter Lysis Buffer
Storage Requirements -20°C